Ethyl palmitate 97

Supplier: Thermo Fisher Scientific

Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester

A15694.14 A15694.22 A15694.36
A15694.14EA 61.8 EUR
A15694.14 A15694.22 A15694.36
Ethyl palmitate 97
Ethyl palmitate

Purity: 97% CAS No.: 628-97-7 Molecular Formula: CH3(CH2)14CO2CH2CH3 Formula Weight: 284.48 Flash Point: 178°(352°F) Refractive Index: 1.4395 Melting Point: 22-26° Boiling Point: 192-193°/10mm Beilstein Reference: 1782663 MDL No.: MFCD00008996

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 192…193 °C (10 mmHg)
Melting Pt: 22…26 °C
Density: 0.857
Flash Pt: 178 °C (352 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008996
CAS Number: 628-97-7
EINECS: 211-064-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR