9-Methylanthracene 99

Supplier: Thermo Fisher Scientific
Danger

A15699.03 A15699.06 A15699.14
A15699.03EA 33.6 EUR
A15699.03 A15699.06 A15699.14
9-Methylanthracene 99
9-Methylanthracene

Purity: 99% CAS No.: 779-02-2 Molecular Formula: C15H12 Formula Weight: 192.26 Melting Point: 77-79° Boiling Point: 196-197°/16mm UN No.: 3077 MDL No.: MFCD00001261

Formula: C₁₅H₁₂
MW: 192.26 g/mol
Boiling Pt: 196…197 °C (16 mmHg)
Melting Pt: 77…79 °C
Density: 1.066
Storage Temperature: Ambient
MDL Number: MFCD00001261
CAS Number: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR