Isophthaloyl dichloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A15904.22 A15904.36
A15904.22EA 37.5 EUR
A15904.22 A15904.36
Isophthaloyl dichloride ≥98%
Isophthaloyl dichloride

Purity: 98% CAS No.: 99-63-8 Molecular Formula: C8H4Cl2O2 Formula Weight: 203.02 Flash Point: 180°(356°F) Melting Point: 42-45° Boiling Point: 275-278° Beilstein Reference: 638342 UN No.: 3261 MDL No.: MFCD00000678

Formula: C₈H₄Cl₂O₂
Boiling Pt: 275…278 °C
Melting Pt: 42…45 °C
Density: 1,388
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000678
CAS Number: 99-63-8
EINECS: 202-774-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR