N-Methylmorpholine N-oxide monohydrate 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Methylmorpholine N-oxide monohydrate, 4-Methylmorpholine 4-oxide hydrate, NMMO

A15996.18 A15996.09 A15996.14 A15996.22
A15996.18EA 130 EUR
A15996.18 A15996.09 A15996.14 A15996.22
N-Methylmorpholine N-oxide monohydrate 98+%
N-Methylmorpholine N-oxide monohydrate

4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.


Purity: 98+% CAS No.: 70187-32-5 Molecular Formula: C5H11NO2.H2O Formula Weight: 135.17 (117.15anhy) Melting Point: 73-76° Beilstein Reference: 5449441

Formula: C₅H₁₁NO₂·1H₂O
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00149388
CAS Number: 70187-32-5
EINECS: 231-391-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR