4-Methoxythiophenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

A16009.06 A16009.14
A16009.06EA 59.8 EUR
A16009.06 A16009.14
4-Methoxythiophenol ≥97%
4-Methoxythiophenol

Purity: 97% CAS No.: 696-63-9 Molecular Formula: C7H8OS Formula Weight: 140.21 Flash Point: 96°(204°F) Refractive Index: 1.5831 Odour: Unpleasant Boiling Point: 100-103°/13mm Beilstein Reference: 1446910 MDL No.: MFCD00004849

Formula: C₇H₈OS
Boiling Pt: 100…103 °C (13 mmHg)
Melting Pt: 42 °C
Density: 1,140
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR