Allyltriphenylphosphonium chloride ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Allyltriphenylphosphonium Chloride

A16012.14 A16012.22 A16012.36
A16012.14EA 48 EUR
A16012.14 A16012.22 A16012.36
Allyltriphenylphosphonium chloride ≥99%
Allyltriphenylphosphonium chloride

Purity: 99% CAS No.: 18480-23-4 Molecular Formula: C21H20ClP Formula Weight: 338.82 Melting Point: 230-232° MDL No.: MFCD00031542

Formula: C₂₁H₂₀ClP
MW: 338.82 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00031542
CAS Number: 18480-23-4
EINECS: 242-368-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR