2'-Deoxyuridine ≥99%

Supplier: Thermo Fisher Scientific

A16026.03 A16026.06 A16026.14
A16026.03EA 26 EUR
A16026.03 A16026.06 A16026.14
2'-Deoxyuridine ≥99%
2'-Deoxyuridine

Purity: 99% CAS No.: 951-78-0 Molecular Formula: C9H12N2O5 Formula Weight: 228.21 Melting Point: 164-167° Beilstein Reference: 24433 Merck Reference: 14,2910 Optical Rotation: +52° (c=1.1 in 1N NaOH) MDL No.: MFCD00006527

Formula: C₉H₁₂N₂O₅
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006527
CAS Number: 951-78-0
EINECS: 213-455-7
Merck Index: 14,2910

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR