TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Supplier: Thermo Fisher Scientific
Warning

A16046.AE A16046.AP A16046.0F
A16046.AEEA 32.2 EUR
A16046.AE A16046.AP A16046.0F
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

CAS No.: 9002-93-1 Formula Weight: ca. 625 Flash Point: 180°(356°F) Physical Form: Liquid Density: 1.061 Refractive Index: 1.4900 Odour: Mild Melting Point: ca 5° Boiling Point: 250° Merck Reference: 14,6761 MDL No.: MFCD00132505


Detergent, equivalent to Nonidet P-40

Boiling Pt: >250 °C
Melting Pt: ∼ 5 °C
Density: 1,061
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 14,6761

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR