2-(2-Butoxyethoxy)ethyl acetate 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethyleneglycolmonobutyletheracetate, Diethylene glycol monobutyl ether acetate

A16065.AK A16065.AU
A16065.AKEA 28.3 EUR
A16065.AK A16065.AU
2-(2-Butoxyethoxy)ethyl acetate 98
2-(2-Butoxyethoxy)ethyl acetate

Purity: 98% CAS No.: 124-17-4 Molecular Formula: CH3CO2CH2CH2OCH2CH2O(CH2)3CH3 Formula Weight: 204.27 Flash Point: 115°(239°F) Physical Form: Liquid Density: 0.977 Refractive Index: 1.4260 Melting Point: -32° Boiling Point: 237-241° Beilstein Reference: 1771533 MDL No.: MFCD00009458

Formula: C₁₀H₂₀O₄
MW: 204.27 g/mol
Boiling Pt: 237…241 °C
Melting Pt: –32 °C
Density: 0.977
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009458
CAS Number: 124-17-4
EINECS: 204-685-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR