1,4-Dichlorobenzene ≥99%

Supplier: Thermo Fisher Scientific
Warning

A16290.0E A16290.22 A16290.36
A16290.0EEA 74.1 EUR
A16290.0E A16290.22 A16290.36
1,4-Dichlorobenzene ≥99%
1,4-Dichlorobenzene

Purity: 99+% CAS No.: 106-46-7 Molecular Formula: C6H4Cl2 Formula Weight: 147.00 Flash Point: 65°(149°F) Odour: Strong Boiling Point: 173-174° Beilstein Reference: 1680023 Merck Reference: 14,3057 UN No.: 3077 MDL No.: MFCD00000604

Formula: C₆H₄Cl₂
Boiling Pt: 173…174 °C
Melting Pt: 52…56 °C
Density: 1,248
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 14,3057

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR