4-(Dimethylamino)butanoic acid hydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

A16610.06 A16610.14
A16610.06EA 56.2 EUR
A16610.06 A16610.14
4-(Dimethylamino)butanoic acid hydrochloride 98%
4-(Dimethylamino)butanoic acid hydrochloride

Purity: 98% CAS No.: 69954-66-1 Molecular Formula: HO2C(CH2)3N(CH3)2.HCl Formula Weight: 167.64 Melting Point: 153-155° MDL No.: MFCD00012613

Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00012613
CAS Number: 69954-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR