Ethyl propionate ≥99%

Supplier: Thermo Fisher Scientific
Danger

A16650.AK A16650.AU
A16650.AKEA 30.5 EUR
A16650.AK A16650.AU
Ethyl propionate ≥99%
Ethyl propionate

Purity: 99% CAS No.: 105-37-3 Molecular Formula: CH3CH2CO2CH2CH3 Formula Weight: 102.13 Flash Point: 12°(54°F) Refractive Index: 1.3850 Melting Point: -74° Boiling Point: 99° Beilstein Reference: 506287 Merck Reference: 14,3847 UN No.: 1195 MDL No.: MFCD00009308

Formula: C₅H₁₀O₂
Boiling Pt: 99 °C
Melting Pt: –74 °C
Density: 0,891
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3
EINECS: 203-291-4
UN: 1195
ADR: 3,II
Merck Index: 14,3847

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR