Sodium chloride 0,9% aqueous solution, sterile

Supplier: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

A1671.0100 A1671.0250 A1671.0500 A1671.1000
A1671.0100EA 74.2 EUR
A1671.0100 A1671.0250 A1671.0500 A1671.1000
Sodium chloride 0,9% aqueous solution, sterile
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

pH (20 °C) 5,0 - 7,5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR