(R)-(+)-4-Benzyl-2-oxazolidinone ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: (R)-4-Benzyl-2-oxazolidinone

A16770.03 A16770.06 A16770.14
A16770.03EA 47.1 EUR
A16770.03 A16770.06 A16770.14
(R)-(+)-4-Benzyl-2-oxazolidinone ≥99%
(R)-(+)-4-Benzyl-2-oxazolidinone

Purity: 99% CAS No.: 102029-44-7 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 86-89° Beilstein Reference: 4782551

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00010846
CAS Number: 102029-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR