1-(2,4-Difluorophenyl)-2-thiourea ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-(2,4-Difluorophenyl)thiourea, 1-(2,4-Difluorophenyl)thiourea, 2,4-Difluorophenylthiourea, (2,4-Difluorophenyl)thiourea

A16945.06
A16945.06EA 0 EUR
A16945.06
1-(2,4-Difluorophenyl)-2-thiourea ≥97%
1-(2,4-Difluorophenyl)-2-thiourea

Purity: 97% CAS No.: 175277-76-6 Molecular Formula: C7H6F2N2S Formula Weight: 188.20 Melting Point: 165-168° UN No.: 2811 MDL No.: MFCD00041150

Formula: (C₆H₃F₂)NC(S)NH₂
MW: 188.2 g/mol
Boiling Pt: 272
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00041150
CAS Number: 175277-76-6
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR