N-(3-(Trimethoxysilyl)propyl)ethylenediamine tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

A16981.0B A16981.18 A16981.30
A16981.0BEA 226 EUR
A16981.0B A16981.18 A16981.30
N-(3-(Trimethoxysilyl)propyl)ethylenediamine tech. 90%, Technical Grade
N-(3-(Trimethoxysilyl)propyl)ethylenediamine

Purity: 90% CAS No.: 1760-24-3 Molecular Formula: (CH3O)3Si(CH2)3NHCH2CH2NH2 Formula Weight: 222.36 Flash Point: 136°(276°F) Refractive Index: 1.4420 Boiling Point: 261-263° Beilstein Reference: 636230 UN No.: 2735 MDL No.: MFCD00008173

Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
Boiling Pt: 261…263 °C
Melting Pt: ∼ 0 °C
Density: 1,010
Flash Pt: 136 °C (276 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008173
CAS Number: 1760-24-3
EINECS: 217-164-6
UN: 2735
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR