CAPS (3-(cyclohexylamino)propanesulphonic acid) ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Cyclohexylaminopropanesulphonic acid

A17037.14 A17037.22 A17037.36
A17037.14EA 39.6 EUR
A17037.14 A17037.22 A17037.36
CAPS (3-(cyclohexylamino)propanesulphonic acid) ≥99%
CAPS (3-(Cyclohexylamino)propanesulphonic acid)

Purity: 99% CAS No.: 1135-40-6 Molecular Formula: C9H19NO3S Formula Weight: 221.32 Odour: Odourless Melting Point: 324° Beilstein Reference: 2835588 Merck Reference: 14,1767 MDL No.: MFCD00003837

Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
Melting Pt: 324
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR