Rose Bengal

Supplier: Thermo Fisher Scientific
Warning

A17053.14 A17053.22 A17053.36
A17053.14EA 47.4 EUR
A17053.14 A17053.22 A17053.36
Rose Bengal
Rose Bengal

CAS No.: 632-69-9 Molecular Formula: C20H2Cl4I4Na2O5 Formula Weight: 1017.65 Melting Point: 300° Beilstein Reference: 3645857 MDL No.: MFCD00151169

Formula: C₂₀H₂Cl₄I₄O₅Na₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9
EINECS: 211-183-3
Merck Index: 14,8262

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR