Azelaic acid tech. 80%, technical

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

A17271.0E A17271.36
A17271.0EEA 125 EUR
A17271.0E A17271.36
Azelaic acid tech. 80%, technical
Azelaic acid

Purity: 80% CAS No.: 123-99-9 Molecular Formula: HOOC(CH2)7COOH Formula Weight: 188.22 Flash Point: 215°(419°F) Melting Point: ca 102° Boiling Point: 286°/100mm Beilstein Reference: 1101094 Merck Reference: 14,905 MDL No.: MFCD00004432

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 286 °C (100 mmHg)
Melting Pt: ∼ 102 °C
Density: 1.225
Flash Pt: 215 °C (419 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Merck Index: 14,905

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR