Isopropyl trifluoroacetate ≥97%

Supplier: Thermo Fisher Scientific
Danger

A17322.14 A17322.22
A17322.14EA 42.6 EUR
A17322.14 A17322.22
Isopropyl trifluoroacetate ≥97%
Isopropyl trifluoroacetate

Purity: 97% CAS No.: 400-38-4 Molecular Formula: CF3CO2CH(CH3)2 Formula Weight: 156.10 Flash Point: -8°(17°F) Refractive Index: 1.3190 Melting Point: -78° Boiling Point: 73° Beilstein Reference: 1764312 UN No.: 2924 MDL No.: MFCD00013566

Formula: C₅H₇F₃O₂
Boiling Pt: 73 °C
Melting Pt: < –78 °C
Density: 1,108
Flash Pt: –8 °C (17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013566
CAS Number: 400-38-4
EINECS: 206-922-1
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR