Metanil yellow

Supplier: Thermo Fisher Scientific
Danger

A17527.18 A17527.30 A17527.PG
A17527.18EA 30.2 EUR
A17527.18 A17527.30 A17527.PG
Metanil yellow
Metanil Yellow

CAS No.: 587-98-4 Molecular Formula: C18H14N3NaO3S Formula Weight: 375.38 Odour: Odourless pH: ~10 Beilstein Reference: 3831568 Merck Reference: 14,5928 MDL No.: MFCD00007486

Formula: C₁₈H₁₄N₃NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007486
CAS Number: 587-98-4
EINECS: 209-608-2
Merck Index: 14,5928

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR