Dithallium trioxide 96%

Supplier: Thermo Fisher Scientific
Danger

A17601.18 A17601.09
A17601.18EA 209 EUR
A17601.18 A17601.09
Dithallium trioxide 96%
Dithallium trioxide

Purity: 96% CAS No.: 1314-32-5 Molecular Formula: Tl2O3 Formula Weight: 456.74 Odour: Odourless Melting Point: 717° Boiling Point: 875° Merck Reference: 14,9268 UN No.: 1707 MDL No.: MFCD00011276

Formula: O₃Tl₂
Boiling Pt: 875 °C
Melting Pt: 717 °C
Density: 9,65
Storage Temperature: Ambient
MDL Number: MFCD00011276
CAS Number: 1314-32-5
EINECS: 215-229-3
UN: 1707
ADR: 6.1,II
Merck Index: 14,9268

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR