2-Methoxynaphthalene 98

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 2-naphthyl ether

A17650.0B A17650.30
A17650.0BEA 119 EUR
A17650.0B A17650.30
2-Methoxynaphthalene 98
2-Methoxynaphthalene

Purity: 98% CAS No.: 93-04-9 Molecular Formula: C11H10O Formula Weight: 158.20 Flash Point: ca 139°(282°F) Melting Point: 70-75° Boiling Point: 272-274° Beilstein Reference: 1859408 Merck Reference: 14,5997 MDL No.: MFCD00004061

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
Boiling Pt: 272…274 °C
Melting Pt: 70…75 °C
Density: 1.266 g/cm³ (20 °C)
Flash Pt: ∼139 °C (282 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004061
CAS Number: 93-04-9
EINECS: 202-213-6
Merck Index: 12,06076

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR