Ethyl (trimethylsilyl)acetate ≥98%

Supplier: Thermo Fisher Scientific
Warning

A17707.09 A17707.18 A17707.30
A17707.09EA 43.4 EUR
A17707.09 A17707.18 A17707.30
Ethyl (trimethylsilyl)acetate ≥98%
Ethyl (trimethylsilyl)acetate

Purity: 98+% CAS No.: 4071-88-9 Molecular Formula: (CH3)3SiCH2CO2CH2CH3 Formula Weight: 160.29 Flash Point: 35°(95°F) Refractive Index: 1.4150 Boiling Point: 156-158° Beilstein Reference: 1755902 UN No.: 3272 MDL No.: MFCD00009172

Formula: C₇H₁₆O₂Si
Boiling Pt: 156…158 °C
Density: 0,876
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009172
CAS Number: 4071-88-9
EINECS: 223-783-2
UN: 3272
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR