Dehydroacetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

A17827.0B A17827.18 A17827.30
A17827.0BEA 189 EUR
A17827.0B A17827.18 A17827.30
Dehydroacetic acid ≥98%
Dehydracetic acid

Purity: 98% CAS No.: 520-45-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Flash Point: 157°(314°F) Melting Point: 109-113° Boiling Point: 269-270° Beilstein Reference: 6129 Merck Reference: 14,2865 MDL No.: MFCD00066709

Formula: C₈H₈O₄
Boiling Pt: 269…270 °C
Melting Pt: 109…113 °C
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 14,2865

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR