4-Bromo-2-chloroaniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

A17859.09 A17859.18 A17859.30
A17859.09EA 49.6 EUR
A17859.09 A17859.18 A17859.30
4-Bromo-2-chloroaniline ≥98%
4-Bromo-2-chloroaniline

Purity: 98+% CAS No.: 38762-41-3 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Melting Point: 68-72° Beilstein Reference: 3117740

Formula: C₆H₅BrClN
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00007660
CAS Number: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR