Squalane ≥98%

Supplier: Thermo Fisher Scientific
Warning

A17931.22 A17931.36
A17931.22EA 60.2 EUR
A17931.22 A17931.36
Squalane ≥98%
Squalane

Purity: 98% CAS No.: 111-01-3 Molecular Formula: C30H62 Formula Weight: 422.83 Flash Point: 218°(424°F) Refractive Index: 1.4510 Melting Point: -38° Boiling Point: 210-215°/1mm Beilstein Reference: 776019 Merck Reference: 14,8767 MDL No.: MFCD00008953

Formula: C₃₀H₆₂
Boiling Pt: 210…215 °C (1 mmHg)
Melting Pt: –38 °C
Density: 0,81
Flash Pt: 218 °C (424 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3
EINECS: 203-825-6
Merck Index: 14,8767

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR