(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)

Supplier: Thermo Fisher Scientific
Warning

A18040.30 A18040.18 A18040.0B
A18040.30EA 75.8 EUR
A18040.30 A18040.18 A18040.0B
(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)
(±)-Camphene

CAS No.: 5794-04-7 Molecular Formula: C10H16 Formula Weight: 136.24 Flash Point: 34°(94°F) Melting Point: ca 35° Boiling Point: 159-160° Merck Reference: 14,1730 UN No.: 1325 MDL No.: MFCD00066603

Formula: C₁₀H₁₆
Boiling Pt: 159…160 °C
Melting Pt: 42…45 °C
Density: 0,850
Flash Pt: 34 °C (94 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066603
CAS Number: 79-92-5
EINECS: 201-234-8
UN: 1325
ADR: 4.1,III
Merck Index: 14,1730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR