4-Fluoro-o-xylene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Fluoro-3,4-dimethylbenzene, 4-Fluoro-1,2-dimethylbenzene

A18253.06 A18253.14 A18253.22
A18253.06EA 49.1 EUR
A18253.06 A18253.14 A18253.22
4-Fluoro-o-xylene ≥98%
4-Fluoro-o-xylene

Purity: 98% CAS No.: 452-64-2 Molecular Formula: C8H9F Formula Weight: 124.16 Flash Point: 40°(104°F) Refractive Index: 1.4815 Odour: Aromatic Boiling Point: 148-149° Beilstein Reference: 1856457 UN No.: 1993 MDL No.: MFCD00039217

Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 148…149 °C
Density: 1.000
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039217
CAS Number: 452-64-2
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR