4-Propylbenzenesulphonyl chloride 95%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Propylbenzene-1-sulphonyl chloride

A18354.14
A18354.14EA 0 EUR
A18354.14
4-Propylbenzenesulphonyl chloride 95%
4-Propylbenzenesulphonyl chloride

Purity: 95% CAS No.: 146949-07-7 Molecular Formula: C9H11ClO2S Formula Weight: 218.70 Refractive Index: 1.5375 Melting Point: 20-22° Boiling Point: 275-276° Beilstein Reference: 5511216 UN No.: 3265 MDL No.: MFCD00041508

Formula: C₉H₁₁ClO₂S
MW: 218,7 g/mol
Boiling Pt: 275…276 °C
Melting Pt: 20…22 °C
Density: 1,223
Storage Temperature: Ambient
MDL Number: MFCD00041508
CAS Number: 146949-07-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR