Pentafluorobenzamide ≥99%

Supplier: Thermo Fisher Scientific
Warning

A18397.06 A18397.14 A18397.22
A18397.06EA 59.5 EUR
A18397.06 A18397.14 A18397.22
Pentafluorobenzamide ≥99%
Pentafluorobenzamide

Purity: 99% CAS No.: 652-31-3 Molecular Formula: C7H2F5NO Formula Weight: 211.09 Melting Point: 146-149° MDL No.: MFCD00007971

Formula: C₇H₂F₅NO
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007971
CAS Number: 652-31-3
EINECS: 211-488-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR