3-Fluoro-p-anisidine ≥98%

Supplier: Thermo Fisher Scientific
Danger

A18606.03 A18606.06 A18606.14
A18606.03EA 42.2 EUR
A18606.03 A18606.06 A18606.14
3-Fluoro-p-anisidine ≥98%
3-Fluoro-p-anisidine

Purity: 98+% CAS No.: 366-99-4 Molecular Formula: C7H8FNO Formula Weight: 141.15 Flash Point: 110°(230°F) Melting Point: 80-84° Boiling Point: 135°/18mm Beilstein Reference: 2717083 UN No.: 2811 MDL No.: MFCD00075040

Formula: C₇H₈FNO
Boiling Pt: 135 °C (18 mmHg)
Melting Pt: 80…84 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075040
CAS Number: 366-99-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR