2-Amino-1-naphthalenesulphonic acid 98%

Supplier: Thermo Fisher Scientific
Warning

A18644.36 A18644.0E
A18644.36EA 63.3 EUR
A18644.36 A18644.0E
2-Amino-1-naphthalenesulphonic acid 98%
2-Amino-1-naphthalenesulphonic acid

Purity: 98% CAS No.: 81-16-3 Molecular Formula: C10H9NO3S Formula Weight: 223.25 Beilstein Reference: 613084 Merck Reference: 14,6406 MDL No.: MFCD00003988

Formula: C₁₀H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00003988
CAS Number: 81-16-3
EINECS: 201-331-5
Merck Index: 14,6406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR