4-Bromo-3,5-dihydroxybenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

A18914.14 A18914.22
A18914.14EA 61.6 EUR
A18914.14 A18914.22
4-Bromo-3,5-dihydroxybenzoic acid ≥98%
4-Bromo-3,5-dihydroxybenzoic acid

Purity: 98+% CAS No.: 16534-12-6 Molecular Formula: C7H5BrO4 Formula Weight: 233.02 Melting Point: 268-272° Beilstein Reference: 2368265

Formula: C₇H₅BrO₄
Melting Pt: 268…272 °C
Storage Temperature: Ambient
MDL Number: MFCD00002513
CAS Number: 16534-12-6
EINECS: 240-606-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR