4-Nitroanisole ≥97%

Supplier: Thermo Fisher Scientific
Warning

A19152.0B A19152.30
A19152.0BEA 341 EUR
A19152.0B A19152.30
4-Nitroanisole ≥97%
4-Nitroanisole

Purity: 97% CAS No.: 100-17-4 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 130°(266°F) Melting Point: 50-54° Boiling Point: 260° Beilstein Reference: 1865361 Merck Reference: 14,6585 UN No.: 3458 MDL No.: MFCD00007327

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 260 °C
Melting Pt: 50…54 °C
Density: 1.233
Flash Pt: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1 III
Merck Index: 12,06682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR