Ethyl-trans-4-decenoate 97%

Supplier: Thermo Fisher Scientific

A19364.09 A19364.18
A19364.09EA 47.4 EUR
A19364.09 A19364.18
Ethyl-trans-4-decenoate 97%
Ethyl-trans-4-decenoate

Purity: 97% CAS No.: 76649-16-6 Molecular Formula: CH3(CH2)4CH=CHCH2CH2CO2CH2CH3 Formula Weight: 198.31 Refractive Index: 1.4370 MDL No.: MFCD00015574

Formula: C₁₂H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00015574
CAS Number: 76649-16-6
EINECS: 278-509-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR