Ethylvanillin ≥98%

Supplier: Thermo Fisher Scientific
Warning

A19478.14 A19478.22 A19478.36
A19478.14EA 27.3 EUR
A19478.14 A19478.22 A19478.36
Ethylvanillin ≥98%
Ethylvanillin

Purity: 98% CAS No.: 121-32-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 127°(260°F) Melting Point: 75-79° Boiling Point: 285° Beilstein Reference: 1073761 Merck Reference: 14,3859 MDL No.: MFCD00006944

Formula: C₉H₁₀O₃
Boiling Pt: 285 °C
Melting Pt: 75…79 °C
Flash Pt: 127 °C (260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 14,3859

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR