4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

A19634.03 A19634.06
A19634.03EA 0 EUR
A19634.03 A19634.06
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥97%
4-Bromo-2,5-difluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 207974-14-9 Molecular Formula: C6H2BrClF2O2S Formula Weight: 291.50 Flash Point: 110°(230°F) Melting Point: 34-36° Boiling Point: 76°/0.01mm UN No.: 3261 MDL No.: MFCD00042181

Formula: C₆H₂BrClF₂O₂S
Boiling Pt: 76 °C (0,01 mmHg)
Melting Pt: 34…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042181
CAS Number: 207974-14-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR