1,3-Diphenyl-2-thiourea ≥98%

Supplier: Thermo Fisher Scientific
Danger

A19644.0E A19644.22 A19644.36
A19644.0EEA 163 EUR
A19644.0E A19644.22 A19644.36
1,3-Diphenyl-2-thiourea ≥98%
1,3-Diphenyl-2-thiourea

Purity: 98% CAS No.: 102-08-9 Molecular Formula: C13H12N2S Formula Weight: 228.32 Melting Point: 150-154° Beilstein Reference: 644277 Merck Reference: 14,3333 UN No.: 2811 MDL No.: MFCD00004921

Formula: C₁₃H₁₂N₂S
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 150…154 °C
Density: 1,320
Storage Temperature: Ambient
MDL Number: MFCD00004921
CAS Number: 102-08-9
EINECS: 203-004-2
UN: 2811
ADR: 6.1,II
Merck Index: 14,3333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR