2,4,6-Trihydroxybenzaldehyde 95%

Supplier: Thermo Fisher Scientific
Warning

A19678.03 A19678.06 A19678.14
A19678.03EA 36.5 EUR
A19678.03 A19678.06 A19678.14
2,4,6-Trihydroxybenzaldehyde 95%
2,4,6-Trihydroxybenzaldehyde

Purity: 95% CAS No.: 487-70-7 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 195° dec. Beilstein Reference: 2254429

Formula: C₇H₆O₄
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: ∼195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003329
CAS Number: 487-70-7
EINECS: 207-663-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR