Tetracycline hydrochloride

Supplier: PanReac AppliChem
Warning

A2228.0025
A2228.0025EA 78.8 EUR
A2228.0025
Tetracycline hydrochloride
Tetracycline hydrochloride
Formula: C₂₂H₂₅ClN₂O₈
MW: 480,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR