Sodium alginate

Supplier: PanReac AppliChem

Synonyms: Alginic acid sodium salt, Algin

A3249.0250 A3249.1000 A3249.9025
A3249.0250EA 110 EUR
A3249.0250 A3249.1000 A3249.9025
Sodium alginate
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR