HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% for molecular biology

Supplier: PanReac AppliChem

Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

A3724.0250 A3724.0500 A3724.1000
A3724.0250EA 214 EUR
A3724.0250 A3724.0500 A3724.1000
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% for molecular biology
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Formula: C₈H₁₈N₂O₄S
Melting Pt: 210…215 °C
Density: 1,448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNases/RNases/Proteases not detectable
Assay min. 99.5 %
pH (1 %; H2O) 4.7 - 6.0
Water max. 0.25 %
Chloride max. 0.005 %
Sulfate max. 0.02 %
As max. 0.0001 %
Fe max. 0.001 %
PB max. 0.001 %
A (1 cm/0.1 M HPLC water) at 260 nm max. 0.1
A (1 cm/0.1 M HPLC water) at 280 nm max. 0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR