Naphthol AS-D chloroacetate

Supplier: PanReac AppliChem

Synonyms: 3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthyl chloroacetate

A4454.0001
A4454.0001EA 155 EUR
A4454.0001
Naphthol AS-D chloroacetate
Naphthol AS-D chloroacetate
Formula: C₂₀H₁₆ClNO₃
MW: 353.8 g/mol
CAS Number: 35245-26-2
EINECS: 252-463-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR