Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Supplier: AAT BIOQUEST
10070
AATB10070EA 537 EUR
AATB10070
Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Assays Enzyme Assays
Malondialdehyde (MDA) is natural byproduct of lipid peroxidation and is widely used as a key indicator to determine the oxidative stress and free radical formation.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.