Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 568)

Supplier: AAT BIOQUEST
100810B0 100810B1
AATB100810B0EA 371 EUR
AATB100810B0 AATB100810B1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 568)
Antibodies
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 568
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR