iFluor® 647 fluorescent probe

Supplier: AAT BIOQUEST
Warning

1011
AATB1011EA 292 EUR
AATB1011
iFluor® 647 fluorescent probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ca19f7f594d8713d6f6aa20553568d96a8aec9b&vid=6ade3efdbcaeded9c3ba83e239066d6ba4ac9557&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR