Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Supplier: AAT BIOQUEST
11111
AATB11111EA 307 EUR
AATB11111
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Assays Protein Assays
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
  • Dynamic range from 12.5 ug/ml to 5 mg/ml of bsa
  • Optimised for cytocite™ and qubit™ fluorometers
  • Highly sensitive
  • Robust protocol
Caution: For research use only.
Order Now
SPECIFICATIONS
Environmentally Preferable