HIS Lite™ Cy3 Bis NTA Chelator
Supplier: AAT BIOQUEST
12650
AATB12650EA 619 EUR
AATB12650
HIS Lite™ Cy3 Bis NTA Chelator
Protein Linking and Labeling Systems
Cy3 Bis NTA-Ni Chelator can be applied either to an SDS-PAGE gel or Western blot membrane for fluorescence imaging or used in living cells in combination with the addition of a heavy metal ion such as Ni²⁺.
  • Stable for 24 months
Quality tested in-house.
Caution: For research use only.