TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Supplier: AAT BIOQUEST
13206
AATB13206EA 965 EUR
AATB13206
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Proteins and Peptides
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2] has much stronger fluoresce upon protease cleavage than the commonly used FRET peptide MCA-HQKLVFFA (K-DNP), a secretase FRET peptide substrate.
  • Stable for 12 months
  • Optimised for FRET applications
Quality tested in-house.
Caution: For research use only.